Sodium acetate, anhydrous, ACS, 99.0% min
Sodium acetate, anhydrous, ACS, 99.0% min
Sodium acetate, anhydrous, ACS, 99.0% min
Sodium acetate, anhydrous, ACS, 99.0% min
Thermo Scientific Chemicals
Sodium acetate, anhydrous, ACS, 99.0% min
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Catalog NumberQuantity
ALF011554.30250 g
Catalog number ALF011554.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium acetate, anhydrous
Melting Point324°C
Name NoteFor ACS analysis
CAS127-09-3
ColorWhite
View more
Sodium acetate is usually used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography and it is also a plating agent.