Cerium(IV) oxide, REacton™, 99.9% (REO)
Cerium(IV) oxide, REacton™, 99.9% (REO)
Cerium(IV) oxide, REacton™, 99.9% (REO)
Cerium(IV) oxide, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Cerium(IV) oxide, REacton™, 99.9% (REO)
CAS: 1306-38-3 | CeO2 | 172.114 g/mol
Have Questions?
Catalog NumberQuantity
ALF011328.30250 g
Catalog number ALF011328.30
Price (MYR)
1,002.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
1,002.00
EA
Add to cart
Specifications
Chemical Name or MaterialCerium(IV) Oxide
Melting Point2600°C
CAS1306-38-3
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Cerium(IV) oxide is used in ceramics, and to polish glass and stones. It is applied as a solid electrolyte in fuel cells. It can also be employed as a co-catalyst in a number of reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass.