Dysprosium(III) oxide, REacton™, 99.9% (REO)
Dysprosium(III) oxide, REacton™, 99.9% (REO)
Dysprosium(III) oxide, REacton™, 99.9% (REO)
Dysprosium(III) oxide, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Dysprosium(III) oxide, REacton™, 99.9% (REO)
For Use in Cermets Used As Nuclear Reaction Control Rods | CAS: 1308-87-8 | Dy2O3 | 373.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF011319.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDysprosium(III) oxide
Melting Point2340°C
CAS1308-87-8
ColorWhite
Recommended StorageAmbient temperatures
View more
In cermets used as nuclear reaction control rodsDysprosium trioxide is used with nickel in cermets, which are used as nuclear reactor control rods. It is also used in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps. It is an important raw material to prepare dysprosium(III) salts by reacting with acids.