Erbium(III) chloride hydrate, REacton™, 99.9% (REO)
Erbium(III) chloride hydrate, REacton™, 99.9% (REO)
Erbium(III) chloride hydrate, REacton™, 99.9% (REO)
Erbium(III) chloride hydrate, REacton™, 99.9% (REO)
Erbium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Erbium(III) chloride hydrate, REacton™, 99.9% (REO)
CAS: 19423-85-9 | Cl3Er | 273.61 g/mol
Have Questions?
Catalog NumberQuantity
ALF011304.22100 g
Catalog number ALF011304.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialErbium(III) chloride hydrate
CAS19423-85-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Erbium(III) chloride hydrate is used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18, for which electron and stability studies were performed.