Europium(III) oxide, REacton™, 99.99% (REO)
Europium(III) oxide, REacton™, 99.99% (REO)
Europium(III) oxide, REacton™, 99.99% (REO)
Europium(III) oxide, REacton™, 99.99% (REO)
Thermo Scientific Chemicals
Europium(III) oxide, REacton™, 99.99% (REO)
CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF011299.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEuropium(III) oxide
Melting Point2291°C
CAS1308-96-9
ColorWhite
Recommended StorageAmbient temperatures
View more
Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.