Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO)
Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO)
Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO)
Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO)
CAS: 13450-84-5 | Cl3GdH12O6 | 371.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF011286.1425 g
Catalog number ALF011286.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGadolinium(III) chloride hexahydrate
Name NoteRare Earth Oxides Content
CAS13450-84-5
ColorWhite
Health Hazard 1H315-H319
View more
Gadolinium(III) chloride hexahydrate is used as a macrophage inhibitor which inhibits the specialized liver macrophages, Kupffer cells that line the walls of the sinusoids.