Neodymium(III) chloride hydrate, REacton™, 99.9% (REO)
Neodymium(III) chloride hydrate, REacton™, 99.9% (REO)
Neodymium(III) chloride hydrate, REacton™, 99.9% (REO)
Neodymium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Neodymium(III) chloride hydrate, REacton™, 99.9% (REO)
CAS: 19423-78-0 | Cl3Nd | 250.59 g/mol
Have Questions?
Catalog NumberQuantity
ALF011251.1850 g
Catalog number ALF011251.18
Price (MYR)
651.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
651.00
EA
Add to cart
Specifications
Chemical Name or MaterialNeodymium(III) chloride hydrate
Melting Point124°C
CAS19423-78-0
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Neodymium(III) chloride hydrate is used for production of neodymium metal. It also finds application in lasers and fiber amplifiers. It serves as dopants for silica based as well as plastic fibers. It plays an important role of additives in organic light emitting diodes resulting in improved LED emission spectrum. In addition, it is used in catalysis of polymerization of various dienes. It is used to modify titanium dioxide for use as a photocatalyst. Its coating on aluminum and its alloy protect them from corrosion. It is also extensively used as fluorescent labels for organic molecules.