Cesium iodide, 99.999% (metals basis)
Cesium iodide, 99.999% (metals basis)
Cesium iodide, 99.999% (metals basis)
Cesium iodide, 99.999% (metals basis)
Thermo Scientific Chemicals
Cesium iodide, 99.999% (metals basis)
CAS: 7789-17-5 | CsI | 259.81 g/mol
Have Questions?
Catalog NumberQuantity
ALF010992.1425 g
Catalog number ALF010992.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point1280°C
Chemical Name or MaterialCesium iodide
Melting Point626°C
CAS7789-17-5
ColorWhite
View more
Cesium iodide is used as an input phosphor in x-ray image intensifier tube of fluoroscopy equipment. It is also used as an equipment for electromagnetic calorimetry in experimental particle physics. It is often employed as a beamsplitter in Fourier Transform Infrared (FT-IR) spectrometers. Cesium iodide is an inorganic scintillator which is mainly used in experimental nuclear physics, high energy physics, nuclear medicine, nuclear tomography, environmental studies and many other fields of use. It is utilized in calorimeters and detectors and also employed as a phosphor in fluoroscopy equipments.