Ytterbium(III) oxide, REacton™, 99.998% (REO)
Ytterbium(III) oxide, REacton™, 99.998% (REO)
Ytterbium(III) oxide, REacton™, 99.998% (REO)
Ytterbium(III) oxide, REacton™, 99.998% (REO)
Thermo Scientific Chemicals
Ytterbium(III) oxide, REacton™, 99.998% (REO)
CAS: 1314-37-0 | O3Yb2 | 394.09 g/mol
Have Questions?
Catalog NumberQuantity
ALF010919.065 g
Catalog number ALF010919.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialYtterbium(III) oxide
Melting Point2227°C
CAS1314-37-0
Recommended StorageAmbient temperatures
Physical FormPowder
View more
It serves as a dopant for silicate mineral crystals in lasers, and a colorant for glasses and enamels. It is employed in the preparation of optical fibers, and special types of alloys. It is used in the preparation of chiral ytterbium complexes which are used as catalysts for enantioselective organic synthesis with high yields. It finds application in optic, electronic and ceramic industries.