Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm
Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm
Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm
Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm
Thermo Scientific Chemicals
Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm
CAS: 12060-58-1
Have Questions?
Catalog NumberQuantity
ALF010915.1425 g
Catalog number ALF010915.14
Price (MYR)
1,776.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,776.00
EA
Add to cart
Specifications
Chemical Name or MaterialSamarium(III) oxide
Melting Point2300°C
Name NoteREM 100ppm
CAS12060-58-1
Recommended StorageAmbient temperatures
View more
Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. It finds use as a neutron absorber in nuclear power reactors to control the reaction. It is also used as a catalyst for the dehydration of primary alcohols to aldehydes or ketones. It forms samarium perchlorate when treated with perchloric acid, which is used as a reference material for checking the wavelength scale of a spectrophotometer in the range of 200 to 500 nm.