trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
Thermo Scientific Chemicals
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
Hydrogenation, hydrosilation, carbonylation, oxidation, and C-C bond formation | CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol
Have Questions?
Catalog NumberQuantity
ALF010491.031 g
Catalog number ALF010491.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-Dichlorobis(triphenylphosphine)palladium(II)
CAS13965-03-2
Melting Point∼310°C (decomposition)
ColorYellow
Recommended StorageAmbient temperatures
View more
As a catalyst of choice in hydrogenation, hydrosilation, carbonylation, oxidation, elimination, reduction, allyl aceate isomerization, dimerization of 1,3-dienes, and C-C bond formation such as Negishi coupling, Suzuki coupling, Kumada coupling, Sonogashira coupling, Heck coupling and Sonogashira-Hagihara coupling reactions. Used in the preparation of tetrakis(triphenylphosphine)palladium, condensed heterocycles such as furoquinolines, diphenylacetylenes, and several other compounds.