Tin powder, -100 mesh, 99.5% (metals basis)
Tin powder, -100 mesh, 99.5% (metals basis)
Tin powder, -100 mesh, 99.5% (metals basis)
Tin powder, -100 mesh, 99.5% (metals basis)
Thermo Scientific Chemicals
Tin powder, -100 mesh, 99.5% (metals basis)
CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Catalog NumberQuantity
ALF010378.22100 g
Catalog number ALF010378.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTin powder
CAS7440-31-5
Health Hazard 1H302-H319-H335-H336-H373-H500
Health Hazard 3P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c