Phosphorus powder, -100 mesh, red amorphous, 98% (metals basis)
Phosphorus powder, -100 mesh, red amorphous, 98% (metals basis)
Phosphorus powder, -100 mesh, red amorphous, 98% (metals basis)
Phosphorus powder, -100 mesh, red amorphous, 98% (metals basis)
Thermo Scientific Chemicals
Phosphorus powder, -100 mesh, red amorphous, 98% (metals basis)
CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF010281.22100 g
Catalog number ALF010281.22
Price (MYR)
469.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
469.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhosphorus powder
CAS7723-14-0
ColorRed to Brown
Physical FormPowder
Quantity100 g