Copper powder, -150 mesh, 99.5% (metals basis)
Copper powder, -150 mesh, 99.5% (metals basis)
Copper powder, -150 mesh, 99.5% (metals basis)
Copper powder, -150 mesh, 99.5% (metals basis)
Thermo Scientific Chemicals
Copper powder, -150 mesh, 99.5% (metals basis)
Apparent density 2.6-2.8g/cm3 | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF010160.36
Price (MYR)
980.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
980.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
Health Hazard 1H228-H315-H319-H335-H500
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c