Chromium powder, -100 mesh, 99% (metals basis)
Chromium powder, -100 mesh, 99% (metals basis)
Chromium powder, -100 mesh, 99% (metals basis)
Chromium powder, -100 mesh, 99% (metals basis)
Thermo Scientific Chemicals
Chromium powder, -100 mesh, 99% (metals basis)
Apparent density 2.0-3.0g/cm3 | CAS: 7440-47-3 | Cr | 52.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Catalog number ALF010147.22
Price (MYR)
506.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
506.00
EA
Add to cart
Specifications
Chemical Name or MaterialChromium
ColorGray
CAS7440-47-3
Health Hazard 1H228
AppearanceGray powder