Bismuth powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Bismuth powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Bismuth powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Bismuth powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bismuth powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF010110.36500 g
Catalog number ALF010110.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBismuth powder
CAS7440-69-9
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228
Flammable solid.
Health Hazard 3P210-P240-P241-P280-P370+P378q