Phosphorus lump, Red, Puratronic™, 99.999+% (metals basis), Thermo Scientific™
Phosphorus lump, Red, Puratronic™, 99.999+% (metals basis), Thermo Scientific™
Phosphorus lump, Red, Puratronic™, 99.999+% (metals basis), Thermo Scientific™
Phosphorus lump, Red, Puratronic™, 99.999+% (metals basis), Thermo Scientific™
Thermo Scientific Chemicals
Phosphorus lump, Red, Puratronic™, 99.999+% (metals basis), Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALF000248.0910 g
Catalog number ALF000248.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhosphorus lump, red
CAS7723-14-0
ColorBrown to Red
Health Hazard 1Danger
Health Hazard 2Hazard statements
H228 Flammable solid
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.