Gold foil, 0.025mm (0.001in) thick, 99.985% (metals basis)
Gold foil, 0.025mm (0.001in) thick, 99.985% (metals basis)
Gold foil, 0.025mm (0.001in) thick, 99.985% (metals basis)
Gold foil, 0.025mm (0.001in) thick, 99.985% (metals basis)
Gold foil, 0.025mm (0.001in) thick, 99.985% (metals basis)
Thermo Scientific Chemicals
Gold foil, 0.025mm (0.001in) thick, 99.985% (metals basis)
Form: Approx.0.3g/25 x 25mm | CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF000133.FF25 x 25 mm
Catalog number ALF000133.FF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialGold
ColorGolden
CAS7440-57-5
AppearanceGold colored foil