Caffeine, citrated
Caffeine, citrated
Caffeine, citrated
Thermo Scientific Chemicals
Caffeine, citrated
Caffeine, citrated, Quantity: 100g, Melting Point: 159.0 deg.C to 161.0 deg.C, Molecular Weight: 386.31, CAS: 69-22-7, Formula Weight: 386.31, Health Hazard 1: GHS Signal Word: Warning, Health Hazard 2: GHS H Statement Harmful if swallowed. Causes serious eye irritation. | CAS: 69-22-7 | C14H18N4O9 | 386.32 g/mol
Have Questions?
Catalog NumberQuantity
ACR46172025025 g
Catalog number ACR461720250
Price (MYR)
963.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
963.00
EA
Add to cart
Specifications
Chemical Name or MaterialCaffeine, citrated
CAS69-22-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.