Cefixime, 98%, Thermo Scientific Chemicals
Cefixime, 98%, Thermo Scientific Chemicals
Cefixime, 98%, Thermo Scientific Chemicals
Cefixime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cefixime, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR4594800505 g
Catalog number ACR459480050
Price (MYR)
1,442.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,442.00
EA
Add to cart
Specifications
Chemical Name or MaterialCefixime
CAS79350-37-1
pH2.6 to 4.1
Infrared SpectrumAuthentic
Assay Percent Range97.5% min. (HPLC)