Gabapentin, 98%
Gabapentin, 98%
Gabapentin, 98%
Thermo Scientific Chemicals
Gabapentin, 98%
Gabapentin, CAS # 60142-96-3, is a synthetic analogue of the neurotransmitter gamma-aminobutyric acid with anticonvulsant and analgesic activities. | CAS: 60142-96-3 | C9H17NO2 | 171.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ACR458020010
Price (MYR)
414.00
Each
Quantity:
1 g
Request bulk or custom format
Price (MYR)
414.00
Each
Specifications
Chemical Name or MaterialGabapentin
Melting Point166.0°C
CAS60142-96-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May damage the unborn child.
View more
General Description
  • Gabapentin is a water-soluble GABA analog with anticonvulsant and analgesic activities
  • The exact mechanism of action is unknown but it has been described that this compound inhibits excitatory neuron activity by interacting with voltage-gated calcium channels in cortical neurons
Application
  • Gabapentin has been used as the α2δ-ligand to examine whether the allyl isothiocyanate (AITC) sensitized thermal aversion assay might be useful as a screening method for the identification of presumptive analgesics
RUO – Research Use Only
General References:
  1. Curtright, A.; Rosser, M.; Goh, S.; Keown, B.; Wagner, E.; Sharifi, J.; Raible, D.W.; Dhaka, A. Modeling nociception in zebrafish: a way forward for unbiased analgesic discovery. PLoS One. 2015, 14, 10, (1), e0116766.
  2. Yasaei, R.; Katta, S.; Saadabadi, A. Gabapentin. [Updated 2021 Dec 27]. In: StatPearls [Internet]. Treasure Island (FL): StatPearls Publishing; 2022.