Lead(II) acetate basic, ACS reagent
Lead(II) acetate basic, ACS reagent
Lead(II) acetate basic, ACS reagent
Lead(II) acetate basic, ACS reagent
Thermo Scientific Chemicals
Lead(II) acetate basic, ACS reagent
CAS: 51404-69-4 | C4H10O8Pb3 | 807.69 g/mol
Have Questions?
Catalog NumberQuantity
ACR4532200101 kg
Catalog number ACR453220010
Price (MYR)
742.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
742.00
EA
Add to cart
Specifications
Chemical Name or MaterialLead(II) acetate basic
CAS51404-69-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Harmful if swallowed. Harmful if inhaled. May damage the unborn child. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Obtain special instructions before use. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray.