Graphene, Single Layer
Graphene, Single Layer
Graphene, Single Layer
Graphene, Single Layer
Graphene, Single Layer
Thermo Scientific Chemicals
Graphene, Single Layer
CAS: 1034343-98-0 | Cx | 16.04 g/mol
Have Questions?
Catalog NumberQuantity
ACR4531500101 g
Catalog number ACR453150010
Price (MYR)
2,754.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
2,754.00
EA
Add to cart
Specifications
Chemical Name or MaterialGraphene
Name NoteSingle Layer
CAS1034343-98-0
Health Hazard 2GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Suitable for battery materials development.
RUO – Research Use Only