Sodium phosphate, tribasic dodecahydrate, extra pure
Sodium phosphate, tribasic dodecahydrate, extra pure
Sodium phosphate, tribasic dodecahydrate, extra pure
Sodium phosphate, tribasic dodecahydrate, extra pure
Thermo Scientific Chemicals
Sodium phosphate, tribasic dodecahydrate, extra pure
CAS: 10101-89-0 | Na3O412H2O | 380.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number ACR450660010
Price (MYR)
453.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
453.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium phosphate, tribasic dodecahydrate
Melting Point75.0°C
CAS10101-89-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement