Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Hydrogen chloride, 3M solution in CPME, AcroSeal™
Thermo Scientific Chemicals
Hydrogen chloride, 3M solution in CPME, AcroSeal™
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR445851000100 mLAcroSeal™ Glass bottle
Catalog number ACR445851000
Price (MYR)
708.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
708.00
EA
Add to cart
Chemical Identifiers
CAS7647-01-0, 5614-37-9
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Boiling Point106.0°C
Health Hazard 1Danger
Density0.8950g/mL