Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR4452600505 g
Catalog number ACR445260050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNorfloxacin
TypeNorfloxacin
Physical FormCrystalline Powder
CAS70458-96-7
Melting Point218°C to 226°C