Water, technical, demineralized
Water, technical, demineralized
Water, technical, demineralized
Water, technical, demineralized
Thermo Scientific Chemicals
Water, technical, demineralized
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR44430025025 LPlastic Can
Catalog number ACR444300250
Price (MYR)
634.00
EA
Add to cart
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Price (MYR)
634.00
EA
Add to cart
Specifications
Chemical Name or MaterialWater
CAS7732-18-5
Boiling Point100.0°C
Molecular FormulaH2O
Linear FormulaH2O