Xylenes, technical
Xylenes, technical
Xylenes, technical
Xylenes, technical
Thermo Scientific Chemicals
Xylenes, technical
CAS: 1330-20-7 | C8H10 | 106.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4442400505 LPlastic Drum
Catalog number ACR444240050
Price (MYR)
545.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
545.00
EA
Add to cart
Specifications
Chemical Name or MaterialXylenes
Melting Point-34.0°C
CAS1330-20-7
Health Hazard 1GHS Signal Word: Danger
Density0.8650g/mL