Ethylene glycol, technical
Ethylene glycol, technical
Ethylene glycol, technical
Ethylene glycol, technical
Thermo Scientific Chemicals
Ethylene glycol, technical
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4442300505 LPlastic Drum
Catalog number ACR444230050
Price (MYR)
622.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
622.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylene Glycol
Melting Point-13.0°C
CAS107-21-1
Density1.1130g/mL
Boiling Point196.0°C to 198.0°C