Acetone, technical
Acetone, technical
Acetone, technical
Thermo Scientific Chemicals
Acetone, technical
CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Catalog NumberQuantity
ACR4441500505 L
Catalog number ACR444150050
Price (MYR)
732.00
EA
Add to cart
Quantity:
5 L
Request bulk or custom format
Price (MYR)
732.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetone
Melting Point-95.0°C
CAS67-64-1
DOT InformationClass 3: Flammable Liquid
Density0.7900g/mL