Clindamycin hydrochloride, 97%
Clindamycin hydrochloride, 97%
Clindamycin hydrochloride, 97%
Clindamycin hydrochloride, 97%
Thermo Scientific Chemicals
Clindamycin hydrochloride, 97%
Clindamycin hydrochloride, CAS # 21462-39-5, is a lincosamide antibiotic. | CAS: 21462-39-5 | C18H34Cl2N2O5S | 461.439 g/mol
Have Questions?
Catalog NumberQuantity
ACR44351010010 mg
Catalog number ACR443510100
Price (MYR)
643.00
EA
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
643.00
EA
Add to cart
Specifications
Chemical Name or MaterialClindamycin hydrochloride
TypeClindamycin Hydrochloride
CAS21462-39-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
View more
General Description
  • Clindamycin is a lincosamide antibiotic derived from lincomycin that has antibiotic activity against aerobic gram-positive cocci, and parasitic protozoa
  • Clindamycin reversibly binds to the 50s bacterial ribosomal subunit, thus inhibiting protein synthesis
Applications
  • Clindamycin has been used to enhance bacterial colonization in research mice during the study of commensal gut microbiota
  • Clindamycin has been used to study biofilm-mediated antibiotic resistance in an in vitro model of G. vaginalis
  • Clindamycin was used as a selection agent to isolate, enumerate, and identify Lactobacillus spp. from probiotic dairy products
RUO – Research Use Only
General References:
  1. Spížek, J.; Řezanka, T. Lincosamides: Chemical structure, biosynthesis, mechanism of action, resistance, and applications. Biochem Pharmacol. 2017, 133:20-28.
  2. Patterson, L.; Allen, J.; Posey, I.; Shaw, J.J.P.; Costa-Pinheiro P, Walker SJ, Gademsey A, Wu X, Wu S, Zachos NC, Fox TE, Sears CL, Kester M. Glucosylceramide production maintains colon integrity in response to Bacteroides fragilis toxin-induced colon epithelial cell signaling. FASEB J. 2020, 34(12), 15922-15945.
  3. Farahmand, N.; Ouoba, L.I.I.; Naghizadeh Raeisi, S.; Sutherland, J.; Ghoddusi, H.B. Probiotic Lactobacilli in Fermented Dairy Products: Selective Detection, Enumeration and Identification Scheme. Microorganisms. 2021, 9(8), 1600.