1,1-Dimethylbiguanide hydrochloride, 97%
1,1-Dimethylbiguanide hydrochloride, 97%
1,1-Dimethylbiguanide hydrochloride, 97%
1,1-Dimethylbiguanide hydrochloride, 97%
Thermo Scientific Chemicals
1,1-Dimethylbiguanide hydrochloride, 97%
CAS: 1115-70-4 | C4H12ClN5 | 165.63 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4297200505 gGlass bottle
Catalog number ACR429720050
Price (MYR)
705.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
705.00
EA
Add to cart
Specifications
Chemical Name or Material1, 1-Dimethylbiguanide hydrochloride
CAS1115-70-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0