Glycogen, From Oyster
Glycogen, From Oyster
Glycogen, From Oyster
Glycogen, From Oyster
Thermo Scientific Chemicals
Glycogen, From Oyster
Glycogen, CAS # 9005-79-2 is a highly branched, polysaccharide form of glucose used for energy storage in humans and other animals. | CAS: 9005-79-2 | C24H42O21 | 666.579 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR422950050
Price (MYR)
1,231.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,231.00
EA
Add to cart
Specifications
Chemical Name or MaterialGlycogen
Name NoteFrom oyster
CAS9005-79-2
ColorBeige to White
RTECS NumberMC2700000
View more
General Description
  • Glycogen is the main form in which energy is stored in animals as well as other higher organisms
  • It consists of a core protein unit surrounded by highly branched glucose units
  • Glycogen is synthesized through a series of enzymatic reactions and stored mainly in the liver and skeletal muscle to provide energy during fasting periods and during muscle contraction
  • Glycogen is an analog of starch, an energy store in plants, though it is both more compact and more highly branched than starch
Applications
  • Glycogen is most commonly used as an inert carrier substance for nucleic acid precipitation that may increase DNA and RNA extraction efficiency
RUO – Research Use Only
General References:
  1. Adeva-Andany, M.M.; González-Lucán, M.; Donapetry-García, C.; Fernández-Fernández, C.; Ameneiros-Rodríguez, E. Glycogen metabolism in humans. BBA Clin. 2016, 5,85-100.
  2. Prykhozhij, S.V.; Steele, S.L.; Razaghi, B.; Berman, J.N. A rapid and effective method for screening, sequencing and reporter verification of engineered frameshift mutations in zebrafish. Dis Model Mech. 2017, 10(6), 811-822.