D-alpha-Tocopheryl acetate, 97%
D-alpha-Tocopheryl acetate, 97%
D-alpha-Tocopheryl acetate, 97%
D-alpha-Tocopheryl acetate, 97%
Thermo Scientific Chemicals
D-alpha-Tocopheryl acetate, 97%
CAS: 58-95-7 | C31H52O3 | 472.75 g/mol
Have Questions?
Catalog NumberQuantity
ACR4210600101 g
Catalog number ACR421060010
Price (MYR)
690.00
BOT
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
690.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD-α-Tocopheryl acetate
Name Note98%
CAS58-95-7
Health Hazard 1PRODUCT NEEDS INERT ATMOSPHERE
ColorYellow