Methyl formate, 98%, for spectroscopy
Methyl formate, 98%, for spectroscopy
Methyl formate, 98%, for spectroscopy
Methyl formate, 98%, for spectroscopy
Methyl formate, 98%, for spectroscopy
Thermo Scientific Chemicals
Methyl formate, 98%, for spectroscopy
CAS: 107-31-3 | C2H4O2 | 60.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR414345000500 mLGlass bottle
Catalog number ACR414345000
Price (MYR)
726.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
726.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl formate
Name Note98%
Acidity0.0008 meq/g max.
CAS107-31-3
Absorbance0.009 max. at 320nm, 0.032 max. at 300nm, 0.051 max. at 270nm, 1 max. at 260nm
View more
Suitable for battery materials development.
for UV/VIS spectroscopy
RUO – Research Use Only