Petroleum ether, extra pure, boiling range 35-60°C
Petroleum ether, extra pure, boiling range 35-60°C
Petroleum ether, extra pure, boiling range 35-60°C
Petroleum ether, extra pure, boiling range 35-60°C
Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 35-60°C
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4131400101 kgGlass Bottle
Catalog number ACR413140010
Price (MYR)
479.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
479.00
EA
Add to cart
Specifications
Chemical Name or MaterialPetroleum ether
CAS64742-49-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.