Check Order Status
alpha-D-Lactose, monohydrate, ACS reagent, ACROS Organics™
Element Search
Structure Search
alpha-D-Lactose, monohydrate, ACS reagent, ACROS Organics™
Have Questions?
Catalog Number
Quantity
Packaging
ACR412975000
500 g
Plastic bottle
Catalog number
ACR412975000
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material
α-D-Lactose, monohydrate
CAS
5989-81-1
CAS
63-42-3
Color
White
Ignition Residue
0.03% max.
View more