Alizarin Yellow R sodium salt, 96%, pure
Alizarin Yellow R sodium salt, 96%, pure
Alizarin Yellow R sodium salt, 96%, pure
Alizarin Yellow R sodium salt, 96%, pure
Thermo Scientific Chemicals
Alizarin Yellow R sodium salt, 96%, pure
CAS: 1718-34-9 | C13H8N3NaO5 | 309.213 g/mol
Have Questions?
Catalog NumberQuantity
ACR40049025025 g
Catalog number ACR400490250
Price (MYR)
511.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
511.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAlizarin Yellow R sodium salt
Name Note96%
CAS1718-34-9
CAS Min %95.0
Health Hazard 1Warning