5-Methoxytryptamine, 97%
5-Methoxytryptamine, 97%
5-Methoxytryptamine, 97%
5-Methoxytryptamine, 97%
Thermo Scientific Chemicals
5-Methoxytryptamine, 97%
CAS: 608-07-1 | C11H14N2O | 190.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3985800101 gGlass bottle
Catalog number ACR398580010
Price (MYR)
923.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
923.00
EA
Add to cart
Specifications
Chemical Name or Material5-Methoxytryptamine
CAS608-07-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi