Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Thermo Scientific Chemicals
Potassium hydroxide, ca. 50% solution in water
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3960600252.5 kgPlastic bottle
Catalog number ACR396060025
Price (MYR)
672.00
BOT
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
672.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium hydroxide
CAS1310-58-3
CAS7732-18-5
Molecular FormulaHKO
Linear FormulaKOH