Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
Thermo Scientific Chemicals
Tetrabutylphosphonium hydroxide, 40 wt.% solution in water
CAS: 14518-69-5 | C16H37OP | 276.45 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR395031000100 mLPlastic Bottle
Catalog number ACR395031000
Price (MYR)
1,266.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,266.00
EA
Add to cart
Chemical Identifiers
CAS14518-69-5, 7732-18-5
Specifications
Chemical Name or MaterialTetrabutylphosphonium hydroxide
CAS14518-69-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
CAS7732-18-5
View more
For C-C Bond Formation (Olefination)
RUO – Research Use Only