L(+)-Tartaric acid, specified according to the requirements of Ph.Eur.
CAS: 87-69-4 | C4H6O6 | 150.09 g/mol
Have Questions?
Catalog Number
Quantity
ACR388280010
1 kg
Catalog number ACR388280010
Price (MYR)
705.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
705.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(+)-Tartaric acid
CAS87-69-4
Melting Point168.0°C to 172.0°C
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes serious eye damage.