Calcium hydroxide, 99.995%, (trace metal basis)
Calcium hydroxide, 99.995%, (trace metal basis)
Calcium hydroxide, 99.995%, (trace metal basis)
Calcium hydroxide, 99.995%, (trace metal basis)
Thermo Scientific Chemicals
Calcium hydroxide, 99.995%, (trace metal basis)
CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3856100505 gGlass bottle
Catalog number ACR385610050
Price (MYR)
804.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
804.00
EA
Add to cart
Specifications
Boiling Point2850.0°C
Chemical Name or MaterialCalcium hydroxide
Melting Point580.0°C
CAS1305-62-0
ColorWhite
View more
for ultra trace analysis
RUO – Research Use Only