Diethylamine, 99.5%, extra pure, redistilled, Thermo Scientific Chemicals
Diethylamine, 99.5%, extra pure, redistilled, Thermo Scientific Chemicals
Diethylamine, 99.5%, extra pure, redistilled, Thermo Scientific Chemicals
Diethylamine, 99.5%, extra pure, redistilled, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethylamine, 99.5%, extra pure, redistilled, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR378371000100 mLGlass bottle
Catalog number ACR378371000
Price (MYR)
730.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
730.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiethylamine
CAS109-89-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre