Isosorbide dimethyl ether, 99%
Isosorbide dimethyl ether, 99%
Isosorbide dimethyl ether, 99%
Isosorbide dimethyl ether, 99%
Thermo Scientific Chemicals
Isosorbide dimethyl ether, 99%
CAS: 5306-85-4 | C8H14O4 | 174.20 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR369911000100 gGlass bottle
Catalog number ACR369911000
Price (MYR)
1,026.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,026.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsosorbide dimethyl ether
CAS5306-85-4
ColorColorless to Yellow
Density1.1700g/mL
Boiling Point93°C to 95°C (0.1 hPa)