4-Formylphenylboronic acid, 97%
4-Formylphenylboronic acid, 97%
4-Formylphenylboronic acid, 97%
4-Formylphenylboronic acid, 97%
Thermo Scientific Chemicals
4-Formylphenylboronic acid, 97%
CAS: 87199-17-5 | C7H7BO3 | 149.94 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR35938025025 gGlass bottle
Catalog number ACR359380250
Price (MYR)
2,554.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,554.00
EA
Add to cart
Specifications
Chemical Name or Material4-Formylphenylboronic acid
Name Note97%
CAS87199-17-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning