Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Thermo Scientific Chemicals
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
CAS: 7291-22-7 | C5H5N | 84.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3514800757.5 mLAmpule
Catalog number ACR351480075
Price (MYR)
1,144.00
PK
Add to cart
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Price (MYR)
1,144.00
PK
Add to cart
Specifications
Chemical Name or MaterialPyridine-d5
Density1.0500g/mL
Deuterium (D)99.75% min.
Flash Point20°C
PackagingAmpule
View more
For NMR
RUO – Research Use Only