Methanol-d4, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.8 atom % D
Methanol-d4, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.8 atom % D
Methanol-d4, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.8 atom % D
Methanol-d4, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.8 atom % D
Thermo Scientific Chemicals
Methanol-d4, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.8 atom % D
CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Catalog NumberQuantity
ACR3514700757.5 mL
Catalog number ACR351470075
Price (MYR)
724.00
BOT
Add to cart
Quantity:
7.5 mL
Request bulk or custom format
Price (MYR)
724.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethanol-d4
Density0.8880g/mL
Deuterium (D)99.75% min.
Flash Point12°C
Physical FormLiquid
View more
For NMR
RUO – Research Use Only