Chitosan, molecular weight: 100,000-300,000
Chitosan, molecular weight: 100,000-300,000
Chitosan, molecular weight: 100,000-300,000
Chitosan, molecular weight: 100,000-300,000
Thermo Scientific Chemicals
Chitosan, molecular weight: 100,000-300,000
Chitosan, CAS # 9012-76-4, is a polymer derived from chitin. It is used in the laboratory research of medical material and biomedicine due to its demonstrated bactericidal, antibacterial, anticancer, and antiviral effects.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
100 g
Packaging:
Plastic bottle
Catalog number ACR349055000
Price (MYR)
3,493.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,493.00
EA
Add to cart
Specifications
Chemical Name or MaterialChitosan
ColorBeige-Yellow to White
Name NoteMolecular Weight: 100, 000 to 300, 000
CAS9012-76-4
Physical FormPowder or Granules
View more
General Description
  • Chitosan is a naturally derived polymer formed from the deacetylation of chitin
  • It is an alkaline polysaccharide with moisturizing and adsorption properties
  • Derivatives of chitosan are produced by chemically modifying chitosan-reactive functional groups
Application
  • Chitosan is used in the laboratory research of medical material and biomedicine due to its bactericidal, antibacterial, anticancer, and antiviral effects
RUO – Research Use Only