Phosphorus, red, 97%, pure, powder
Phosphorus, red, 97%, pure, powder
Thermo Scientific Chemicals
Phosphorus, red, 97%, pure, powder
CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Catalog NumberQuantity
ACR345975000500 g
Catalog number ACR345975000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhosphorus, red
Melting Point590.0°C
CAS7723-14-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Flammable solid.
Harmful to aquatic life with long lasting effects.